Merck
  • Gold-catalyzed carboalkoxylations of 2-ethynylbenzyl ethers to form 1- and 3-substituted 2-methoxy-1-H-indenes: Brønsted acids versus gold catalysis.
Gold-catalyzed carboalkoxylations of 2-ethynylbenzyl ethers to form 1- and 3-substituted 2-methoxy-1-H-indenes: Brønsted acids versus gold catalysis.
Organic & biomolecular chemistry (2014-10-31)
Chun-Hao Chen, Chiou-Dong Wang, Yi-Feng Hsieh, Rai-Shung Liu
ABSTRAKT
Selective synthesis of 1- and 3-substituted 2-methoxyindenes from the carboalkoxylations of 2-ethynylbenzyl ethers is described; the former is obtained efficiently with P(t-Bu)2(o-biphenyl)AuCl/NaBARF in DCM/MS 4 Å whereas the latter is produced preferably with P(t-Bu)2(o-biphenyl)AuCl/AgNTf2 in pre-dried DCM. Both 1- and 3-substituted 2-indenyl ethers are subjected to ozone oxidations to afford two distinct carbonyl products. Our new data indicate that 1-substituted 2-indenyl ethers are generated from gold catalysts whereas their 3-substituted analogues arise from Brønsted acids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS