Merck
Synthesis of sFlt-1 by platelet-monocyte aggregates contributes to the pathogenesis of preeclampsia.
American journal of obstetrics and gynecology (2014-01-21)
Heather D Major, Robert A Campbell, Robert M Silver, D Ware Branch, Andrew S Weyrich
ABSTRAKT
Soluble fms-like tyrosine kinase (sFlt-1) is an important mediator in the pathogenesis of preeclampsia. We sought to determine whether platelet-monocyte aggregates (PMAs) produced sFlt-1 and whether PMAs contributed to sFlt-1 production in preeclampsia. This was a case-control study of sFlt-1 release from PMAs using blood samples from women with preeclampsia matched by gestational age to pregnant controls. A third group of nonpregnant, reproductive-age women comprised an additional control group. Experiments were also performed using blood from nonpregnant women to elucidate whether inducing PMAs could stimulate sFlt-1 production and, if so, to determine the necessary receptors and pathways. Women with preeclampsia had increased total Flt-1 concentrations in platelets and monocytes at baseline compared with pregnant controls (25 vs 10 pg/mL, P = .0003). sFlt-1 production was elicited from monocytes incubated with thrombin-activated platelets from nonpregnant women. sFlt-1 production was regulated at the transcriptional level by p38 and nuclear factor-κB-dependent pathways. Activated platelets in preeclampsia bind monocytes to generate sFlt-1. PMAs are a previously unrecognized source of sFlt-1 that may contribute to endothelial dysfunction and systemic inflammation commonly observed in preeclampsia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture