Merck
Quantitative selenium speciation by HPLC-ICP-MS(IDA) and simultaneous activity measurements in human vitreous humor.
Analytical and bioanalytical chemistry (2014-10-27)
Raquel González de Vega, María Luisa Fernández-Sánchez, Héctor González Iglesias, Miguel Coca Prados, Alfredo Sanz-Medel
ABSTRAKT
A post-column isotope dilution analysis (IDA) methodology was applied to carry out quantitative speciation of selenium in human vitreous humor samples by size exclusion chromatography (SEC) coupled on-line to inductively coupled plasma mass spectrometry (ICP-MS). Two main selenium species detected by SEC-ICP-MS were found to be associated to protein complexes. The expected molecular weights for both selenium-bound complexes were confirmed by MALDI-TOF(MS) and the results matched well with the theoretical mass of a GPx monomer (M, 22 kDa) and tetramer (T, 88 kDa). The quantification of the two detected selenium-bound complexes by post-column IDA showed that the total content of selenospecies in vitreous humor was approximately 3.2 ± 1.8 ppb Se. Moreover, in most of the analyzed vitreous humor samples, the majority of the selenium was associated to higher molecular weight GPx biomolecules. In an attempt to assess if the enzymatic activity was associated with a given selenium-bound GPx protein, the antioxidant enzyme activity was assayed for the two separated GPx species. Only for GPx (T) was a linear relationship between activity and total Se concentration found by ICP-MS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%