Merck
Ab initio structure search and in situ 7Li NMR studies of discharge products in the Li-S battery system.
Journal of the American Chemical Society (2014-11-11)
Kimberly A See, Michal Leskes, John M Griffin, Sylvia Britto, Peter D Matthews, Alexandra Emly, Anton Van der Ven, Dominic S Wright, Andrew J Morris, Clare P Grey, Ram Seshadri
ABSTRAKT
The high theoretical gravimetric capacity of the Li-S battery system makes it an attractive candidate for numerous energy storage applications. In practice, cell performance is plagued by low practical capacity and poor cycling. In an effort to explore the mechanism of the discharge with the goal of better understanding performance, we examine the Li-S phase diagram using computational techniques and complement this with an in situ (7)Li NMR study of the cell during discharge. Both the computational and experimental studies are consistent with the suggestion that the only solid product formed in the cell is Li2S, formed soon after cell discharge is initiated. In situ NMR spectroscopy also allows the direct observation of soluble Li(+)-species during cell discharge; species that are known to be highly detrimental to capacity retention. We suggest that during the first discharge plateau, S is reduced to soluble polysulfide species concurrently with the formation of a solid component (Li2S) which forms near the beginning of the first plateau, in the cell configuration studied here. The NMR data suggest that the second plateau is defined by the reduction of the residual soluble species to solid product (Li2S). A ternary diagram is presented to rationalize the phases observed with NMR during the discharge pathway and provide thermodynamic underpinnings for the shape of the discharge profile as a function of cell composition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Supelco
1,2-Dimethoxyethane, analytical standard
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
1,2-Dimethoxyethane, suitable for HPLC, 99.9%, inhibitor-free
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Węgiel - Szklisty, rod, 200mm, diameter 5.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 7.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, foil, 100x100mm, thickness 2.0mm, glassy carbon
Węgiel - Szklisty, rod, 200mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 50mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 6.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, rod, 200mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foil, 10x10mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Węgiel - Szklisty, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%