Merck
Protective effect of Lactobacillus plantarum 21, a probiotic on trinitrobenzenesulfonic acid-induced ulcerative colitis in rats.
International immunopharmacology (2015-03-03)
C S V Satish Kumar, K Kondal Reddy, A Gopala Reddy, A Vinoth, Sudha Rani Chowdary Ch, G Boobalan, G S Rao
ABSTRAKT
Inflammatory mediators play a crucial role in ulcerative colitis (UC). The present study was aimed to evaluate the effects of Lactobacillus plantarum 21 (LAB 21) on inflammatory mediators in trinitrobenzenesulfonic acid (TNBS)-induced colitis in rats. The inflammatory response was assessed by changes in colon morphology, histopathology, and measurement of reduced glutathione (GSH), lipid peroxidation (TBARS), nitric oxide (NO), interleukin 1β (IL-1β), tumor necrosis factor α (TNF-α), and interleukin 10 (IL-10) mRNA and protein levels by ELISA. Besides, protein expressions of IL-1β and IL-10 were also evaluated by western blot. Treatment with LAB 21 (1×10(10)CFU/rat/day) and sulfasalazine (500mgkg(-1) body weight) for 14days after induction of colitis, significantly decreased TBARS, NO and increased GSH concentration. The protein and mRNA expressions of IL-1β and TNFα were down-regulated, whereas, protein and mRNA expression of IL-10 was up-regulated in LAB 21-treated rats. Moreover, LAB 21 attenuated the macroscopic colonic damage, histopathological changes induced by TNBS. These results suggest that LAB 21 may be effective in the treatment of UC by immunomodulatory and antioxidant properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Picrylsulfonic acid solution, 1 M in H2O