Merck
Direct and stereospecific interaction of amphidinol 3 with sterol in lipid bilayers.
Biochemistry (2014-04-30)
Rafael Atillo Espiritu, Nobuaki Matsumori, Masashi Tsuda, Michio Murata
ABSTRAKT
Amphidinol 3 (AM3), a polyhydroxy-polyene metabolite from the dinoflagellate Amphidinium klebsii, possesses potent antifungal activity. Although AM3 permeabilizes phospholipid membranes only in the presence of sterol, the detailed molecular basis by which AM3 recognizes sterols in membranes remains unknown. Here, we investigated the molecular interaction between sterols and AM3 in membranes from the viewpoint of stereospecific molecular recognition using ergosterol, cholesterol, and epicholesterol, which is the 3-OH epimer of cholesterol. Dye leakage assays, surface plasmon resonance experiments, (2)H and (31)P NMR measurements, and microscopic observations revealed that AM3 directly interacts with membrane sterols through the strict molecular recognition of the stereochemistry of the sterol 3-OH group. The direct interaction enhances the membrane binding efficiency of AM3, which subsequently permeabilizes membranes without altering membrane integrity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.