Merck
  • Proteomic analysis of the sheep caruncular and intercaruncular endometrium reveals changes in functional proteins crucial for the establishment of pregnancy.
Proteomic analysis of the sheep caruncular and intercaruncular endometrium reveals changes in functional proteins crucial for the establishment of pregnancy.
Reproduction (Cambridge, England) (2014-01-22)
K H Al-Gubory, M Arianmanesh, C Garrel, S Bhattacharya, P Cash, P A Fowler
ABSTRAKT
The expression and regulation of endometrial proteins are crucial for conceptus implantation and development. However, little is known about site-specific proteome profiles of the mammalian endometrium during the peri-implantation period. We utilised a two-dimensional gel electrophoresis/mass spectrometry-based proteomics approach to compare and identify differentially expressed proteins in sheep endometrium. Caruncular and intercaruncular endometrium were collected on days 12 (C12) and 16 (C16) of the oestrous cycle and at three stages of pregnancy corresponding to conceptus pre-attachment (P12), implantation (P16) and post-implantation (P20). Abundance and localisation changes in differentially expressed proteins were determined by western blot and immunohistochemistry. In caruncular endometrium, 45 protein spots (5% of total spots) altered between day 12 of pregnancy (P12) and P16 while 85 protein spots (10% of total spots) were differentially expressed between P16 and C16. In intercaruncular endometrium, 31 protein spots (2% of total spots) were different between P12 and P16 while 44 protein spots (4% of total spots) showed differential expression between C12 and C16. The pattern of protein changes between caruncle and intercaruncle sites was markedly different. Among the protein spots with implantation-related changes in volume, 11 proteins in the caruncular endometrium and six proteins in the intercaruncular endometrium, with different functions such as protein synthesis and degradation, antioxidant defence, cell structural integrity, adhesion and signal transduction, were identified. Our findings highlight the different but important roles of the caruncular and intercaruncular proteins during early pregnancy.