Merck
The performance of a small-calibre graft for vascular reconstructions in a senescent sheep model.
Biomaterials (2014-08-12)
Maqsood Ahmed, George Hamilton, Alexander M Seifalian
ABSTRAKT
There is an acute clinical need for small-calibre (<6 mm) vascular grafts for surgery. The aim of this study was to evaluate the long-term performance of a small-calibre graft produced from a nanocomposite biomaterial, polyhedral oligomeric silsesquioxane poly(carbonate-urea)urethane (POSS-PCU), in a large animal model following Good Manufacturing Practice (GMP) and Good Laboratory Practice (GLP) protocols. Grafts were characterised and implanted into the left carotid artery (LCA) of senescent sheep (n = 11) for a period of 9 months. In vivo compliance and blood flow rates were measured using ultrasound wall tracking software and a Transonic flow meter. Graft patency and degree of intimal hyperplasia (IH) were examined at the study end point. Seven of the POSS-PCU grafts were free from thrombosis, IH, calcification and aneurysmal dilation, with 4 occluding within 14 days. All of the ePTFE controls (n = 4) were found to be occluded by day 32. The lumen of the patent POSS-PCU grafts was free from any cellular deposits, whilst perigraft tissue could be seen to be infiltrating into the body of the graft from the adventitia. No significant differences were detected between the blood flow rates (p = 0.3693) and compliance (p = 0.9706) of the POSS-PCU grafts and the native artery, either post-operatively or after 9 months implantation. Small-calibre vascular grafts produced from POSS-PCU offer a viable option for the clinical use in revascularisation procedures with a patency rate of 64%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Supelco
N,O-Bis(trimethylsilyl)acetamide, derivatization grade (GC derivatization), LiChropur, ≥98.5% (GC)
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Węgiel - Szklisty, rod, 200mm, diameter 5.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 7.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, foil, 100x100mm, thickness 2.0mm, glassy carbon
Węgiel - Szklisty, rod, 200mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 50mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 6.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, rod, 200mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foil, 10x10mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Węgiel - Szklisty, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%