Merck
Clinical implications of species identification in monomicrobial Aeromonas bacteremia.
PloS one (2015-02-14)
Chi-Jung Wu, Po-Lin Chen, Po-Ren Hsueh, Ming-Chung Chang, Pei-Jane Tsai, Hsin-I Shih, Hsuan-Chen Wang, Pei-Hsin Chou, Wen-Chien Ko
ABSTRAKT
Advances in Aeromonas taxonomy have led to the reclassification of aeromonads. Hereon, we aimed to re-evaluate the characteristics of Aeromonas bacteremia, including those of a novel species, Aeromonas dhakensis. A retrospective study of monomicrobial Aeromonas bacteremia at a medical center in southern Taiwan from 2004-2011 was conducted. Species identification was based on rpoB sequencing. Of bacteremia of 153 eligible patients, A. veronii (50 isolates, 32.7%), A. dhakensis (48, 31.4%), A. caviae (43, 28.1%), and A. hydrophila (10, 6.5%) were the principal causative species. A. dhakensis and A. veronii bacteremia were mainly community-acquired and presented as primary bacteremia, spontaneous bacterial peritonitis, or skin and soft-tissue infection, whereas A. caviae was associated with hospital-onset bacteremia. The distribution of the AmpC β-lactamase and metallo-β-lactamase genes was species-specific: bla(AQU-1), bla(MOX), or bla(CepH) was present in A. dhakensis, A. caviae, or A. hydrophila, respectively, and bla(CphA) was present in A. veronii, A. dhakensis, and A. hydrophila. The cefotaxime resistance rates of the A. caviae, A. dhakensis, and A. hydrophila isolates were higher than that of A. veronii (39.5%%, 25.0%, and 30% vs. 2%, respectively). A. dhakensis bacteremia was linked to the highest 14-day sepsis-related mortality rate, followed by A. hydrophila, A. veronii, and A. caviae bacteremia (25.5%, 22.2%, 14.0%, and 4.7%, respectively; P = 0.048). Multivariate analysis revealed that A. dhakensis bacteremia, active malignancies, and a Pitt bacteremia score ≥ 4 was an independent mortality risk factor. Characteristics of Aeromonas bacteremia vary between species. A. dhakensis prevalence and its associated poor outcomes suggest it an important human pathogen.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Digoxigenin, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard