Merck
  • Synthesis of aminoacylated N(6),N(6)-dimethyladenosine solid support for efficient access to hydrolysis-resistant 3'-charged tRNA mimics.
Synthesis of aminoacylated N(6),N(6)-dimethyladenosine solid support for efficient access to hydrolysis-resistant 3'-charged tRNA mimics.
Bioorganic & medicinal chemistry (2014-12-03)
Sandro Neuner, Ronald Micura
ABSTRAKT
RNA-amino acid and RNA-peptide conjugates that mimic charged tRNA 3'-ends are valuable substrates for structural and functional investigations of ribosomal complexes. To obtain such conjugates, most synthetic approaches that are found in the literature make use of puromycin. This well available aminonucleoside antibiotic contains a dimethylamino group at the nucleobase and a methylated tyrosine that is connected via an amide linkage to the ribose moiety. To increase structural diversity, we present the synthesis of a N(6),N(6)-dimethylated 3'-azido-3'-deoxyadenosine precursor that can be coupled to any amino acid. Further derivatization results in the solid support that is eligible for the preparation of stable 3'-aminoacyl- or 3'-peptidyl-tRNA termini with an amide instead of the natural ester linkage. The present work expands our previously established route that delivered a broad range of peptidyl-tRNA mimics to the corresponding counterparts with N(6),N(6)-dimethylation pattern of the terminal adenosine (A76). This aspect is of significance to modulate the binding preferences of the mimics for ribosomal A- versus P-site.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%