Merck
  • Determination of benzodiazepines, related pharmaceuticals and metabolites in water by solid-phase extraction and liquid-chromatography-tandem mass spectrometry.
Determination of benzodiazepines, related pharmaceuticals and metabolites in water by solid-phase extraction and liquid-chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-06-14)
Inés Racamonde, Rosario Rodil, José Benito Quintana, Eugenia Villaverde-de-Sáa, Rafael Cela
ABSTRAKT
This work presents a method for the simultaneous determination of 23 benzodiazepines, metabolites and related pharmaceuticals (zolpidem, methylphenidate and ritalinic acid) by solid-phase extraction (SPE) followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS). Different SPE cartridges were considered: hydrophilic modified reversed-phase (Oasis HLB) and their modified versions containing also a cationic-exchange group (Oasis MCX) or anionic-exchange (Oasis MAX) funcionalities. Stability of analytes and the impact of the final eluate volume on the matrix effects on LC-MS/MS were also considered. In the final method, 100-200 mL of sample are extracted with Oasis MCX (60 mg), eluted with 5 mL of methanol (1.25% NH3) and the eluate concentrated and analyzed by LC-MS/MS. Under these conditions, LOQs were established between 0.1 and 18 ng L(-1) for influent wastewater. The use of surrogated deuterated internal standards allows obtaining recoveries in the 84-122% range. Finally the method was applied to determinate the analytes in wastewater and surface water and 10 compounds were detected in the range of 0.5-170 ng L(-1), being the ritalinic acid (the main metabolite of methylphenidate) the analyte detected in the highest concentrations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Clonazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Nordiazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Prazepam
Supelco
Temazepam, analytical standard
Flunitrazepam, European Pharmacopoeia (EP) Reference Standard
Supelco
Flunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Desmethyldiazepam, analytical standard
Clonazepam, European Pharmacopoeia (EP) Reference Standard