Merck
Comparison of the biological properties of ProRoot MTA, OrthoMTA, and Endocem MTA cements.
Journal of endodontics (2014-07-24)
Miri Kim, Wonkyung Yang, Heesun Kim, Hyunjung Ko
ABSTRAKT
OrthoMTA (BioMTA, Seoul, Korea) and Endocem MTA (Maruchi, Wonju-si, Korea) were recently developed to overcome the disadvantages of ProRoot MTA (Dentsply, Tulsa, OK). This study aimed to compare the biological properties of OrthoMTA and Endocem MTA with those of ProRoot MTA using the preosteoblastlike cell line MC3T3-E1. The setting times of calcium silicate-based cements (CSCs) and their effects on the pH of distilled water during storage were determined according to ISO standards. MC3T3-E1 cells were cultured with ProRoot MTA, OrthoMTA, and Endocem MTA. The viability of the cells was assessed using the Cell Counting Kit-8 assay (Dojindo Laboratory, Kumamoto, Japan) on the supernatants of CSCs, and the cells' osteopontin production was determined by an enzyme-linked immunosorbent assay on a culture with the materials on days 3 and 7 of incubation. Endocem MTA exhibited a significantly shorter setting time (15.3 ± 0.5 minutes) than did ProRoot MTA and OrthoMTA (318.0 ± 56.0 and 324.3 ± 2.1 minutes, P < .05). Additionally, all CSCs caused their storage water to become highly alkaline after 7 days. OrthoMTA was significantly more cytotoxic than ProRoot and Endocem MTA (P < .05). ProRoot MTA induced significantly more OPN production than OrthoMTA and Endocem MTA on both days 3 and 7 (P < .05). ProRoot MTA appeared to be superior to OrthoMTA and Endocem MTA in terms of biological properties although Endocem MTA exhibited the shortest setting time and presented lower cytotoxicity with osteoblastlike cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Wodorowęglan sodu, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%