Merck
Enzymatic resolution of ibuprofen in an organic solvent under ultrasound irradiation.
Biotechnology and applied biochemistry (2014-04-05)
Dantong Zhao, Hong Yue, Ge Chen, Liyan Jiang, Hong Zhang, Zhi Wang, Guangchun Liu
ABSTRAKT
Ultrasound has been successfully adopted to improve the biocatalytic properties of APE1547 (a novel esterase from the archaeon Aeropyrum pernix K1) in the resolution of ibuprofen. After optimizing the conditions (ultrasound power, 200 W; temperature, 35 °C), the best biocatalytic performance of APE1547 (enzyme activity, 5.39 µmol/H/mg; E value, 130.8) was obtained. Compared with the conventional reaction in an orbital shaker, the enzyme activity was significantly enhanced about 90-fold, and the enantioselectivity was enhanced about fourfold after an ultrasound. The results of scanning electron microscopy clearly indicated that the activation effect of ultrasound on APE1547 originated mainly in the morphological change of the enzyme powder. Both lower particle size and conformational change of APE1547 under ultrasound might be helpful to enhance the enantioselectivity. In addition, APE1547 kept its best performance under the low-power ultrasound for at least five reaction cycles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard