Merck
Expression of inflammatory markers in a genetic rodent model of depression.
Behavioural brain research (2014-10-04)
Nina Strenn, Petra Suchankova, Staffan Nilsson, Christina Fischer, Gregers Wegener, Aleksander A Mathé, Agneta Ekman
ABSTRAKT
The complex bidirectional communication between the central nervous system and the peripheral immune system is of possible relevance for both normal brain functions and the development of psychiatric disorders. The aim of this investigation was to study central expression of inflammatory markers in a genetic rat model of depression (the Flinders sensitive line (FSL) and its control, the Flinders resistant line (FRL)). A peripheral immune activation was induced by lipopolysaccharide (LPS) in order to investigate possible differences in immune reactions between the two rat lines. To confirm behavioural differences between the rat lines the forced swim test was performed, a test to assess depressive-like behaviour. Expression of candidate inflammatory genes was measured in amygdala, hippocampus, hypothalamus, prefrontal cortex and striatum using quantitative real time PCR. Our results show, for the first time, significantly lower central expression of the glial-specific protein S100B and complement factor C3 in several brain regions of the FSL rats compared to controls, both at baseline and after peripheral immune stimulation. No significant differences in immune responses to LPS were observed between the rats lines. Both S100B and C3 have been suggested to be of relevance for brain development and plasticity as well as brain disorders. These proteins may be of importance for the behavioural differences between the FSL and FRL rats, and this model may be useful in studies exploring the influence of the immune system on brain functions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Oxytocin