Merck
Adaptor protein DOK3 promotes plasma cell differentiation by regulating the expression of programmed cell death 1 ligands.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-24)
Xijun Ou, Shengli Xu, Yan-Feng Li, Kong-Peng Lam
ABSTRAKT
The adaptor Downstream-of-Kinase (DOK) 3 functions as a negative regulator and attenuates B-cell receptor-mediated calcium signaling. Although DOK3 is dispensable for early B-cell development, its role in plasma cell (PC) differentiation is unknown. Here, we show that Dok3(-/-) mice have increased populations of T follicular-helper (Tfh) and germinal center (GC) B cells upon immunization with a T-cell-dependent antigen. However, interestingly, they generate significantly fewer PCs. Bone marrow reconstitution experiments show that the PC defect is B-cell intrinsic and due to the inability of Dok3(-/-) B cells to sustain programmed cell death 1 (PD-1) ligand 1 (PDL1) and up-regulate PD-1 ligand 2 (PDL2) expressions that are critical for PC differentiation. Overexpression of PDL2 rectifies the PC differentiation defect in Dok3(-/-) B cells. We further demonstrate that calcium signaling suppresses the transcription of PD-1 ligands. Abrogation of calcium signaling in B cells by deleting BTK or PLCγ2 or inhibiting calcineurin with cyclosporine A leads to increased expression of PD-1 ligands. Thus, our study reveals DOK3 as a nonredundant regulator of PC differentiation by up-regulating PD-1 ligand expression through the attenuation of calcium signaling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%