Merck
Pentraxin 3 is a local inflammatory marker in atrial fibrillation.
Heart and vessels (2013-08-28)
Takeshi Soeki, Sachiko Bando, Etsuko Uematsu, Tomomi Matsuura, Toshiyuki Niki, Takayuki Ise, Kenya Kusunose, Junko Hotchi, Yuka Ueda, Noriko Tomita, Koji Yamaguchi, Shusuke Yagi, Daiju Fukuda, Yoshio Taketani, Takashi Iwase, Hirotsugu Yamada, Tetsuzo Wakatsuki, Michio Shimabukuro, Masataka Sata
ABSTRAKT
Increasing evidence indicates that inflammation contributes to the pathogenesis of atrial fibrillation (AF). Pentraxin 3 (PTX3) is produced abundantly in local inflammatory lesions while C-reactive protein (CRP) is produced mainly in the liver. In this study, we investigated whether a local level of PTX3 might be a sensitive marker for the local inflammation of AF. Blood from the periphery and left atrial appendage (LAA) was sampled from 23 patients with AF undergoing pulmonary vein isolation, and from 10 control subjects with Wolff-Parkinson-White syndrome. We measured peripheral and LAA plasma concentrations of CRP, interleukin-6 (IL-6), tumor necrosis factor-α (TNF-α), and PTX3. Plasma PTX3 concentrations in both locations were higher in patients with AF than in control subjects. PTX3 concentrations were significantly higher in the LAA than the periphery in patients with AF (3.7 ± 1.4 vs 3.3 ± 1.2 ng/ml, P < 0.01), but not in control subjects (2.4 ± 0.5 vs 2.4 ± 0.5 ng/ml, not significant). Patients and controls showed no significant differences in CRP, IL-6, or TNF-α concentrations between the periphery and LAA. Interestingly, there was a significant positive correlation between LAA plasma concentrations of PTX3 and left atrial volume (r = 0.55, P < 0.01). These data demonstrate that local PTX3 production in the left atrium might reflect the local inflammation of AF.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-PITX3 antibody produced in rabbit, affinity isolated antibody