Merck
Cardiac autonomic modulation of children with Down syndrome.
Pediatric cardiology (2014-08-29)
Tatiana Dias de Carvalho, Luiz Carlos de Abreu, Zan Mustacchi, Luiz Carlos Marques Vanderlei, Moacir Fernandes Godoy, Rodrigo Daminello Raimundo, Celso Ferreira Filho, Talita Dias da Silva, Laura Guilhoto, Viviane Perico, Vivian Ribeiro Finotti, Celso Ferreira
ABSTRAKT
The aim of this study is to analyze the autonomic modulation in children with Down syndrome (DS). The study was conducted with a convenience sample of children with DS and without heart disease, from the Genetics Clinic of the Hospital Infantil Darcy Vargas and APAE São Paulo, São Paulo, SP, Brazil. The control group was matched for sex and age. The analysis of autonomic modulation was performed using the indices of heart rate variability (HRV). The children remained in the supine position with spontaneous breathing for 20 min. Heart rate was recorded beat-to-beat. HRV analysis was performed in time and frequency domain. For data analysis, we used Student's t test: unpaired and Mann-Whitney. It was considered statistically significant at p < 0.05. From 75 children with DS, 50 were excluded, a total of 25 children [16 boys, 8.6 (1.4) years] participated in this study, and the control group also consisted of 25 children [16 boys, 9.0 (1.2) years] without the syndrome. The BMI of the volunteers with DS was higher than the controls [19.1 (2.9) vs. 15.8 (1.2), p < 0.0001]. There were differences between groups in the indices in frequency domain: LFms(2) [1242.1 (788.25) vs. 786.44 (481.90), p = 0.040], LFun [69.104 (11.247) vs. 57.348 (11.683), p = 0.0004], HFun [30.896 (11.247) vs. 42.520 (11.634), p = 0.0004] and LF/HF [2.594 (1.104) vs. 1.579 (0.9982), p = 0.0004]. No differences were observed in time domain indices. The results indicate increased indices representing the sympathetic branch of the autonomic nervous system and those that indicate the overall modulation in children with DS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis