Merck
Injectable gel scaffold based on biopolymer microspheres via an enzymatic reaction.
Advanced healthcare materials (2014-05-17)
Huaping Tan, Ming Fan, Ye Ma, Jiacai Qiu, Xianming Li, Jingxuan Yan
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BMP-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
4-Maleimidobutyric acid N-hydroxysuccinimide ester, ≥98.0% (HPLC)
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Human BMP2 ELISA Kit, for serum, plasma, cell culture supernatant and urine