Merck
Propofol modulates phasic and tonic GABAergic currents in spinal ventral horn interneurones.
British journal of anaesthesia (2014-08-26)
V S Eckle, U Rudolph, B Antkowiak, C Grasshoff
ABSTRAKT
Surgical interventions like skin incisions trigger withdrawal reflexes which require motor neurones and local circuit interneurones in the spinal ventral horn. This region plays a key role in mediating immobilizing properties of the GABAergic anaesthetic propofol. However, it is unclear how propofol modulates GABA(A) receptors in the spinal ventral horn and whether tonic or phasic inhibition is involved. Organotypic spinal cord tissue slices were prepared from mice. Whole-cell recordings were performed for quantifying effects of propofol on GABA(A) receptor-mediated phasic transmission and tonic conductance. Propofol increased GABAergic phasic transmission by a prolongation of the decay time constant in a concentration-dependent manner. The amount of the charge transferred per inhibitory post-synaptic current, described by the area under the curve, was significantly augmented by 1 µM propofol (P<0.01). A GABA(A) receptor-mediated tonic current was not induced by 1 µM propofol but at a concentration of 5 µM (P<0.05). Propofol depresses ventral horn interneurones predominantly by phasic rather than by tonic GABA(A) receptor-mediated inhibition. However, the present results suggest that the involvement of a tonic inhibition might contribute to the efficacy of propofol to depress nociceptive reflexes at high concentrations of the anaesthetic.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard