Merck
Pharmacological protection of retinal pigmented epithelial cells by sulindac involves PPAR-α.
Proceedings of the National Academy of Sciences of the United States of America (2014-11-12)
Arunodoy Sur, Shailaja Kesaraju, Howard Prentice, Kasirajan Ayyanathan, Diane Baronas-Lowell, Danhong Zhu, David R Hinton, Janet Blanks, Herbert Weissbach
ABSTRAKT
The retinal pigmented epithelial (RPE) layer is one of the major ocular tissues affected by oxidative stress and is known to play an important role in the etiology of age-related macular degeneration (AMD), the major cause of blinding in the elderly. In the present study, sulindac, a nonsteroidal antiinflammatory drug (NSAID), was tested for protection against oxidative stress-induced damage in an established RPE cell line (ARPE-19). Besides its established antiinflammatory activity, sulindac has previously been shown to protect cardiac tissue against ischemia/reperfusion damage, although the exact mechanism was not elucidated. As shown here, sulindac can also protect RPE cells from chemical oxidative damage or UV light by initiating a protective mechanism similar to what is observed in ischemic preconditioning (IPC) response. The mechanism of protection appears to be triggered by reactive oxygen species (ROS) and involves known IPC signaling components such as PKG and PKC epsilon in addition to the mitochondrial ATP-sensitive K(+) channel. Sulindac induced iNOS and Hsp70, late-phase IPC markers in the RPE cells. A unique feature of the sulindac protective response is that it involves activation of the peroxisome proliferator-activated receptor alpha (PPAR-α). We have also used low-passage human fetal RPE and polarized primary fetal RPE cells to validate the basic observation that sulindac can protect retinal cells against oxidative stress. These findings indicate a mechanism for preventing oxidative stress in RPE cells and suggest that sulindac could be used therapeutically for slowing the progression of AMD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Fenofibrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Fenofibrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sulindac, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Sulindac, ≥98.0%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
USP
Fenofibrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fenofibrate, ≥99%, powder