Merck
Determination of preservatives in cosmetics, cleaning agents and pharmaceuticals using fast liquid chromatography.
Journal of chromatographic science (2013-01-18)
Irena Baranowska, Iwona Wojciechowska, Natalia Solarz, Ewa Krutysza
ABSTRAKT
This paper reports the development of a method for simultaneously determining five preservatives in cosmetics, cleaning agents and pharmaceuticals by fast liquid chromatography. Methylisothiazolinone, methylchloroisothiazolinone, benzyl alcohol, sodium benzoate and methylparaben were separated on a Chromolith Fast Gradient reversed-phase 18e column using gradient elution with acetonitrile and a 0.1% aqueous solution of formic acid, with a run time of 3 min. The preparation of solid and liquid samples included ultrasonic extraction with methanol with recoveries ranging from 69 to 119%. The developed method was used to analyze samples of cosmetics (66 samples), cleaning agents (five samples) and pharmaceutical industry products (17 samples).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Metyloparaben, United States Pharmacopeia (USP) Reference Standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Supelco
2-Methyl-4-isothiazolin-3-one, analytical standard
Sigma-Aldrich
2-Methyl-4-isothiazolin-3-one, 95%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)