Merck
Snail1 correlates with patient outcomes in E-cadherin-preserved gastroesophageal junction adenocarcinoma.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2013-12-21)
H Dong, L Xie, C Tang, S Chen, Q Liu, Q Zhang, W Zheng, Z Zheng, H Zhang
ABSTRAKT
The poor prognosis of gastroesophageal junction (GEJ) adenocarcinoma is largely associated with metastasis. We here report the first study to investigate the expression of epithelial-mesenchymal transition (EMT) markers Snail1 and E-cadherin in GEJ adenocarcinoma. Snail1 and E-cadherin were detected by immunohistochemistry in a cohort of 128 patients with surgically resected GEJ adenocarcinoma. We assessed the pathologic and prognostic relevance in all patients and within clinically different preserved E-cadherin and reduced E-cadherin-expressing sub-groups. Immunoreactivity for Snail1 and E-cadherin was positive in 68 and 43 % of tumors, respectively. Snail1-positive tumors had more frequent lymph node metastasis and advanced tumor stage. E-cadherin expression was highly associated with histological differentiation, tumor size, advanced stage, presence of lymph node metastasis and distant metastasis. Patients with positive E-cadherin expression or negative Snail1 expression had significantly favorable overall survival rate. In E-cadherin-preserved tumors, the expression of Snail1 was related to lymph node metastasis, advanced stage and poor patient outcome. However, Snail1 expression had no statistically significant relationship with clinicopathologic parameters or prognosis in the reduced E-cadherin-expressing sub-group. Multivariate survival analysis identified that tumor stage [hazard ratio (HR) 2.440; 95 % confidence interval (CI) 1.216-4.896; P = 0.012], lymph node metastasis (HR 2.404; 95 % CI 1.188-4.867; P = 0.015) and gender (HR 3.244; 95 % CI 1.568-6.714; P = 0.002) were independent prognostic markers for overall survival. Snail1 may act more critically in E-cadherin-positive tumors. Evaluation of Snail1 and E-cadherin in GEJ adenocarcinoma may help in assessing malignant properties and stratifying patients.