Merck
  • Effects of the traditional Chinese medicine Yi Shen Jian Gu granules on aromatase inhibitor-associated musculoskeletal symptoms: a study protocol for a multicenter, randomized, controlled clinical trial.
Effects of the traditional Chinese medicine Yi Shen Jian Gu granules on aromatase inhibitor-associated musculoskeletal symptoms: a study protocol for a multicenter, randomized, controlled clinical trial.
Trials (2014-06-03)
Nan Peng, Yi Zhang, Cong Ma, Ming-Wei Yu, Guo-Wang Yang, Qi Fu, Wei-Ru Xu, Xiao-Min Wang
ABSTRAKT
Aromatase inhibitors (AIs) are widely used as an adjuvant endocrine treatment in postmenopausal women with early-stage breast cancer. One of the main adverse effects of AIs is musculoskeletal symptoms, which leads to a lower quality of life and poor adherence to AI treatment. To date, no effective management of aromatase inhibitor-associated musculoskeletal symptoms (AIMSS) has been developed. To determine whether the traditional Chinese medicine Yi Shen Jian Gu granules could effectively manage AIMSS we will conduct a multicenter, randomized, double-blind, placebo-controlled clinical trial. Patients experiencing musculoskeletal symptoms after taking AIs will be enrolled and treated with traditional Chinese medicine or placebo for 12 weeks. The primary outcome measures include Brief Pain Inventory-Short Form, Western Ontario and McMaster Universities Osteoarthritis Index, and Modified Score for the Assessment and Quantification of Chronic Rheumatoid Affections of the Hands, which will be obtained at baseline and at 4, 8, 12 and 24 weeks. The results of this study will provide a new strategy to help relieve AIMSS. ISRCTN06129599 (assigned 14 August 2013).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Letrozole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Letrozole, ≥98% (HPLC)
Letrozole, European Pharmacopoeia (EP) Reference Standard