Merck
Radical-cation salts of BEDT-TTF with lithium tris(oxalato)metallate(III).
Dalton transactions (Cambridge, England : 2003) (2015-03-05)
Lee Martin, Hendrik Engelkamp, Hiroki Akutsu, Shin'ichi Nakatsuji, Jun'ichi Yamada, Peter Horton, Michael B Hursthouse
ABSTRAKT
The first radical-cation salts in the extensive family (BEDT-TTF)x[(A)M(C2O4)3]·Guest containing lithium as the counter cation have been synthesized and characterised.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Carvone, ≥97%, FCC, FG
Sigma-Aldrich
Bis(ethylenedithio)tetrathiafulvalene, 98%
Sigma-Aldrich
L-Carvone, natural, 99%, FG
Sigma-Aldrich
12-Crown-4, 98%
Sigma-Aldrich
(R)-(−)-Carvone, 98%
Supelco
(−)-Carvone, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%