- Synthesis and anti-HIV activity of GS-9148 (2'-Fd4AP), a novel nucleoside phosphonate HIV reverse transcriptase inhibitor.
Synthesis and anti-HIV activity of GS-9148 (2'-Fd4AP), a novel nucleoside phosphonate HIV reverse transcriptase inhibitor.
Bioorganic & medicinal chemistry letters (2008-01-01)
Constantine G Boojamra, Richard L Mackman, David Y Markevitch, Vidya Prasad, Adrian S Ray, Janet Douglas, Deborah Grant, Choung U Kim, Tomas Cihlar
PMID18164198
ABSTRAKT
GS-9148 (2'-Fd4AP, 4) has been identified as a nucleoside phosphonate reverse transcriptase (RT) inhibitor with activity against wild-type HIV (EC(50)=12 microM). Unlike many clinical RT inhibitors, relevant reverse transcriptase mutants (M184V, K65R, 6-TAMs) maintain a susceptibility to 2'-Fd4AP that is similar to wild-type virus. The 2'-fluorine group was rationally designed into the molecule to improve the selectivity profile and in preliminary studies using HepG2 cells, compound 4 showed no measurable effect on mitochondrial DNA content indicating a low potential for mitochondrial toxicity.