Merck
Warfarin glycosylation invokes a switch from anticoagulant to anticancer activity.
ChemMedChem (2011-06-30)
Pauline Peltier-Pain, Shannon C Timmons, Agnès Grandemange, Etienne Benoit, Jon S Thorson
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Deoxy-L-ribose, ≥97.0% (TLC)