Merck
  • Transformations of monoterpenoids in aqueous acids: The reactions of linalool. geraniol, nerol and their acetates in aqueous citric acid.
Transformations of monoterpenoids in aqueous acids: The reactions of linalool. geraniol, nerol and their acetates in aqueous citric acid.
Tetrahedron (1978)
Baxter RL ,et al.