Merck
ETV4 and Myeov knockdown impairs colon cancer cell line proliferation and invasion.
Biochemical and biophysical research communications (2006-05-09)
Alan C Moss, Garrett Lawlor, David Murray, Dónal Tighe, Stephen F Madden, Anne-Marie Mulligan, Conor O Keane, Hugh R Brady, Peter P Doran, Padraic MacMathuna
ABSTRAKT
We have identified novel colorectal cancer-associated genes using NCBI's UNIGENE cDNA libraries. Colon cancer libraries were examined using Digital Differential Display and disease-associated genes were selected. Among these were ETV4 and MYEOV, novel colorectal cancer-associated genes. Samples of matched normal and neoplastic colon were obtained from human subjects and gene expression was quantified using real-time PCR. ETV4 gene expression was significantly increased in colonic neoplasia in comparison to matched normal colonic tissue (p<0.05). Myeov expression was also increased in colon neoplasia in comparison to matched normal tissue. The effect of siRNA-mediated knockdown of ETV4 and Myeov on cell proliferation and invasion was assessed. ETV4 knockdown resulted in a 90% decrease in cell proliferation (p<0.05) and a 67% decrease in cell invasion. Myeov knockdown resulted in a 48% decrease in cell proliferation (p<0.05) and a 36% decrease in cell invasion. These data suggest that ETV4 and Myeov may provide novel targets for therapeutic intervention.