Merck
A missense mutation in KCTD17 causes autosomal dominant myoclonus-dystonia.
American journal of human genetics (2015-05-20)
Niccolo E Mencacci, Ignacio Rubio-Agusti, Anselm Zdebik, Friedrich Asmus, Marthe H R Ludtmann, Mina Ryten, Vincent Plagnol, Ann-Kathrin Hauser, Sara Bandres-Ciga, Conceição Bettencourt, Paola Forabosco, Deborah Hughes, Marc M P Soutar, Kathryn Peall, Huw R Morris, Daniah Trabzuni, Mehmet Tekman, Horia C Stanescu, Robert Kleta, Miryam Carecchio, Giovanna Zorzi, Nardo Nardocci, Barbara Garavaglia, Ebba Lohmann, Anne Weissbach, Christine Klein, John Hardy, Alan M Pittman, Thomas Foltynie, Andrey Y Abramov, Thomas Gasser, Kailash P Bhatia, Nicholas W Wood
ABSTRAKT
Myoclonus-dystonia (M-D) is a rare movement disorder characterized by a combination of non-epileptic myoclonic jerks and dystonia. SGCE mutations represent a major cause for familial M-D being responsible for 30%-50% of cases. After excluding SGCE mutations, we identified through a combination of linkage analysis and whole-exome sequencing KCTD17 c.434 G>A p.(Arg145His) as the only segregating variant in a dominant British pedigree with seven subjects affected by M-D. A subsequent screening in a cohort of M-D cases without mutations in SGCE revealed the same KCTD17 variant in a German family. The clinical presentation of the KCTD17-mutated cases was distinct from the phenotype usually observed in M-D due to SGCE mutations. All cases initially presented with mild myoclonus affecting the upper limbs. Dystonia showed a progressive course, with increasing severity of symptoms and spreading from the cranio-cervical region to other sites. KCTD17 is abundantly expressed in all brain regions with the highest expression in the putamen. Weighted gene co-expression network analysis, based on mRNA expression profile of brain samples from neuropathologically healthy individuals, showed that KCTD17 is part of a putamen gene network, which is significantly enriched for dystonia genes. Functional annotation of the network showed an over-representation of genes involved in post-synaptic dopaminergic transmission. Functional studies in mutation bearing fibroblasts demonstrated abnormalities in endoplasmic reticulum-dependent calcium signaling. In conclusion, we demonstrate that the KCTD17 c.434 G>A p.(Arg145His) mutation causes autosomal dominant M-D. Further functional studies are warranted to further characterize the nature of KCTD17 contribution to the molecular pathogenesis of M-D.