Merck
Transamidation of primary amides with amines using hydroxylamine hydrochloride as an inorganic catalyst.
Angewandte Chemie (International ed. in English) (2012-01-04)
C Liana Allen, Benjamin N Atkinson, Jonathan M J Williams
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%