Merck
  • Simple and efficient synthesis of 3-aminopropenones and 3-aminopropenoates catalyzed by copper (II) nitrate trihydrate under solvent-free conditions.
Simple and efficient synthesis of 3-aminopropenones and 3-aminopropenoates catalyzed by copper (II) nitrate trihydrate under solvent-free conditions.
Monatshefte fur Chemie / Chemical Monthly (2008)
Li GC.