Merck
A deleterious mutation in SAMD9 causes normophosphatemic familial tumoral calcinosis.
American journal of human genetics (2006-09-09)
Orit Topaz, Margarita Indelman, Ilana Chefetz, Dan Geiger, Aryeh Metzker, Yoram Altschuler, Mordechai Choder, Dani Bercovich, Jouni Uitto, Reuven Bergman, Gabriele Richard, Eli Sprecher
ABSTRAKT
Familial tumoral calcinosis (FTC) is a rare autosomal recessive disorder characterized by the progressive deposition of calcified masses in cutaneous and subcutaneous tissues, which results in painful ulcerative lesions and severe skin and bone infections. Two major types of FTC have been recognized: hyperphosphatemic FTC (HFTC) and normophosphatemic FTC (NFTC). HFTC was recently shown to result from mutations in two different genes: GALNT3, which codes for a glycosyltransferase, and FGF23, which codes for a potent phosphaturic protein. To determine the molecular cause of NFTC, we performed homozygosity mapping in five affected families of Jewish Yemenite origin and mapped NFTC to 7q21-7q21.3. Mutation analysis revealed a homozygous mutation in the SAMD9 gene (K1495E), which was found to segregate with the disease in all families and to interfere with the protein expression. Our data suggest that SAMD9 is involved in the regulation of extraosseous calcification, a process of considerable importance in a wide range of diseases as common as atherosclerosis and autoimmune disorders.