Merck
  • Associations between autoimmune thyroid disease prognosis and functional polymorphisms of susceptibility genes, CTLA4, PTPN22, CD40, FCRL3, and ZFAT, previously revealed in genome-wide association studies.
Associations between autoimmune thyroid disease prognosis and functional polymorphisms of susceptibility genes, CTLA4, PTPN22, CD40, FCRL3, and ZFAT, previously revealed in genome-wide association studies.
Journal of clinical immunology (2012-06-19)
Naoya Inoue, Mikio Watanabe, Hiroya Yamada, Kazuya Takemura, Fumiaki Hayashi, Noriko Yamakawa, Maiko Akahane, Yu Shimizuishi, Yoh Hidaka, Yoshinori Iwatani
ABSTRAKT
Genome-wide association studies have revealed several susceptibility genes among patients with autoimmune thyroid disease (AITD), including CTLA4, PTPN22, FCRL3, and ZFAT. However, any possible association between these genes and AITD prognosis remains unknown. The objective of this study was to identify associations between polymorphisms of these genes and AITD prognosis. We genotyped functional polymorphisms, including CTLA4 CT60, CTLA4 +49A/G, CTLA4 -1147C/T, CTLA4 -318C/T, PTPN22 -1123C/G, PTPN22 SNP37, CD40 -1C/T, FCRL3 -169C/T, ZFAT Ex9b-SNP10, and ZFAT Ex9b-SNP2, in 197 AITD patients carefully selected from 456 registered AITD patients, and 86 control subjects. The restriction fragment length polymorphism method was used for genotyping. The CD40 -1CC genotype and C allele were significantly more frequent in patients with Graves' disease (GD) in remission than in those with intractable GD (P = 0.041 and P = 0.031, respectively). The FCRL3 -169TT genotype was significantly less frequent in patients with intractable GD than in those with GD in remission (P = 0.0324). For a ZFAT Ex9b-SNP10 polymorphism, the TT genotype and T allele were significantly more frequent in patients with severe Hashimoto's disease (HD) than in those with mild HD (P = 0.0029 and P = 0.0049, respectively). For a CTLA4 CT60 polymorphism, the antithyrotropin receptor antibody levels at the onset of GD were significantly higher in those with the GG genotype than in those with other genotypes (P = 0.0117). CD40 and FCRL3 gene polymorphisms were associated with GD intractability, and ZFAT polymorphism was associated with HD severity but not its development.