Merck
  • A novel chordin-like BMP inhibitor, CHL2, expressed preferentially in chondrocytes of developing cartilage and osteoarthritic joint cartilage.
A novel chordin-like BMP inhibitor, CHL2, expressed preferentially in chondrocytes of developing cartilage and osteoarthritic joint cartilage.
Development (Cambridge, England) (2003-12-09)
Naoki Nakayama, Chun-ya E Han, Linh Cam, Jae I Lee, Jim Pretorius, Seth Fisher, Robert Rosenfeld, Sheila Scully, Ryuichi Nishinakamura, Diane Duryea, Gwyneth Van, Brad Bolon, Takashi Yokota, Ke Zhang
ABSTRAKT
We have identified a novel chordin-like protein, CHL2, which is structurally most homologous to CHL/neuralin/ventroptin. When injected into Xenopus embryos, CHL2 RNA induced a secondary axis. Recombinant CHL2 protein interacted directly with BMPs in a competitive manner to prevent binding to the type I BMP receptor ectodomain, and inhibited BMP-dependent induction of alkaline phosphatase in C2C12 cells. Thus, CHL2 behaves as a secreted BMP-binding inhibitor. In situ hybridization revealed that CHL2 expression is restricted to chondrocytes of various developing joint cartilage surfaces and connective tissues in reproductive organs. Adult mesenchymal progenitor cells expressed CHL2, and its levels decreased during chondrogenic differentiation. Addition of CHL2 protein to a chondrogenic culture system reduced cartilage matrix deposition. Consistently, CHL2 transcripts were weakly detected in normal adult joint cartilage. However, CHL2 expression was upregulated in middle zone chondrocytes in osteoarthritic joint cartilage (where hypertrophic markers are induced). CHL2 depressed chondrocyte mineralization when added during the hypertrophic differentiation of cultured hyaline cartilage particles. Thus, CHL2 may play negative roles in the (re)generation and maturation of articular chondrocytes in the hyaline cartilage of both developing and degenerated joints.