Merck
  • Vapor pressure of acetophenone,(?)-1,2-butanediol,(?)-1,3-butanediol, diethylene glycol monopropyl ether, 1,3-dimethyladamantane, 2-ethoxyethyl acetate, ethyl octyl sulfide, and pentyl acetate.
Vapor pressure of acetophenone,(?)-1,2-butanediol,(?)-1,3-butanediol, diethylene glycol monopropyl ether, 1,3-dimethyladamantane, 2-ethoxyethyl acetate, ethyl octyl sulfide, and pentyl acetate.
Journal of Chemical and Engineering Data (1996)
Steele WV, et al.