- Intestinal amebae.
ABSTRAKT
Among the Entamoeba species that infect humans, Entamoeba histolytica causes diseases, Entamoeba dispar is a harmless commensal, Entamoeba moshkovskii seems to be a pathogen, and the pathogenicity of Entamoeba bangladeshi remains to be investigated. Species-specific detection needed for treatment decisions and for understanding the epidemiology and pathogenicity of these amebae. Antigen-based detection methods are needed for E dispar, E moshkovskii, and E bangladeshi; and molecular diagnostic test capable of detecting E histolytica, E dispar, E moshkovskii, and E bangladeshi simultaneously in clinical samples. Next-generation sequencing of DNA from stool is needed to identify novel species of Entamoeba.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)