Merck
  • Simple synthesis of biocompatible biotinylated porous hexagonal ZnO nanodisc for targeted doxorubicin delivery against breast cancer cell: In vitro and in vivo cytotoxic potential.
Simple synthesis of biocompatible biotinylated porous hexagonal ZnO nanodisc for targeted doxorubicin delivery against breast cancer cell: In vitro and in vivo cytotoxic potential.
Colloids and surfaces. B, Biointerfaces (2015-06-21)
Prasun Patra, Shouvik Mitra, Amarto Das Gupta, Saheli Pradhan, Saurav Bhattacharya, Manisha Ahir, Sudeshna Mukherjee, Sampad Sarkar, Subhrodeb Roy, Sreya Chattopadhyay, Arghya Adhikary, Arunava Goswami, Dhrubajyoti Chattopadhyay
ABSTRAKT
Targeted drug delivery with porous materials features great promise as improved therapeutic potential for treatment of various diseases. In the present study we have attempted a microwave synthesis of porous hexagonal nanodisc of zinc oxide (PZHD) for the first time and its subsequent targeted delivery to breast cancer cells, MCF7. PZHD has been fabricated suitably with 3-aminopropyltriethoxysilane to impart additional stability and surface amines to anchor site directing ligand NHS-biotin. Biotinylated scaffold showed targeted delivery of anticancer drug doxorubicin and pH triggered release to MCF 7 cells with preferential distribution on specified domain. A detailed in vitro cytotoxicity study was associated with it to evaluate the mode of action of Dox loaded PZHD on MCF-7 cells by means of cell cycle analysis, apoptosis assays, Western blot and immuno-fluorescence image analysis. The efficacy of the Dox loaded PZHD was further validated from our in vivo tumor regression studies. Finally, the whole study has been supported by in vitro and in vivo bio-safety studies which also signified its biocompatibility with real time applications. To the best of our knowledge this is the first effort to use biotinylated PZHD for targeted delivery of doxorubicin within MCF 7 cells with a detailed study of its mechanistic application. This study might thus hold future prospects for therapeutic intervention for treatment of cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)