Merck
Results of (68)Gallium-DOTATATE PET/CT Scanning in Patients with Multiple Endocrine Neoplasia Type 1.
Journal of the American College of Surgeons (2015-07-25)
Samira M Sadowski, Corina Millo, Candice Cottle-Delisle, Roxanne Merkel, Lily A Yang, Peter Herscovitch, Karel Pacak, William F Simonds, Stephen J Marx, Electron Kebebew
ABSTRAKT
Screening for neuroendocrine tumors (NETs) in patients with multiple endocrine neoplasia type 1 (MEN1) is recommended to detect primary and metastatic tumors, which can result in significant morbidity and mortality. The utility of somatostatin receptor imaging (68)Gallium-DOTATATE PET/CT in patients with MEN1 is not known. The aim of this study was to prospectively determine the accuracy of (68)Gallium-DOTATATE PET/CT vs (111)In- pentetreotide single-photon emission CT (SPECT)/CT and anatomic imaging in patients with MEN1. We performed a prospective study comparing (68)Gallium-DOTATATE PET/CT, (111)In-pentetreotide SPECT/CT, and triphasic CT scan to clinical, biochemical, and pathologic data in 26 patients with MEN1. (68)Gallium-DOTATATE PET/CT detected 107 lesions; (111)In-pentetreotide SPECT/CT detected 33 lesions; and CT scan detected 48 lesions. Lesions detected on (68)Gallium-DOTATATE PET/CT had high standard uptake value (SUV)(max) (median SUV(max) = 72.8 [range 19 to 191]). In 7 of the 26 patients (27%), (68)Gallium-DOTATATE PET/CT was positive, with a negative (111)In-pentetreotide SPECT/CT, and in 10 patients (38.5%), additional metastases were detected (range 0.3 cm to 1.5 cm). In 8 of the 26 patients (31%), there was a change in management recommendations as a result of the findings on (68)Gallium-DOTATATE PET/CT that were not seen on (111)In-pentetreotide SPECT/CT and CT scan. (68)Gallium-DOTATATE PET/CT is more sensitive for detecting NETs than (111)In-pentetreotide SPECT/CT and CT scan in patients with MEN1. This imaging technique should be integrated into radiologic screening and surveillance of patients with MEN1 because it can significantly alter management recommendations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems