Merck
Differences in inhibitory avoidance, cortisol and brain gene expression in TL and AB zebrafish.
Genes, brain, and behavior (2015-04-25)
M Gorissen, R Manuel, T N M Pelgrim, W Mes, M J S de Wolf, J Zethof, G Flik, R van den Bos
ABSTRAKT
Recently, we established an inhibitory avoidance paradigm in Tupfel Long-Fin (TL) zebrafish. Here, we compared task performance of TL fish and fish from the AB strain; another widely used strain and shown to differ genetically and behaviourally from TL fish. Whole-body cortisol and telencephalic gene expression related to stress, anxiety and fear were measured before and 2 h post-task. Inhibitory avoidance was assessed in a 3-day paradigm: fish learn to avoid swimming from a white to a black compartment where a 3V-shock is given: day 1 (first shock), day 2 (second shock) and day 3 (no shock, sampling). Tupfel Long-Fin fish rapidly learned to avoid the black compartment and showed an increase in avoidance-related spatial behaviour in the white compartment across days. In contrast, AB fish showed no inhibitory avoidance learning. AB fish had higher basal cortisol levels and expression levels of stress-axis related genes than TL fish. Tupfel Long-Fin fish showed post-task learning-related changes in cortisol and gene expression levels, but these responses were not seen in AB fish. We conclude that AB fish show higher cortisol levels and no inhibitory avoidance than TL fish. The differential learning responses of these Danio strains may unmask genetically defined risks for stress-related disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
DL-Dithiothreitol solution, 1 M in H2O