Merck
  • Comparative studies on the inhibitory activities of selected benzoic acid derivatives against secretory phospholipase A2, a key enzyme involved in the inflammatory pathway.
Comparative studies on the inhibitory activities of selected benzoic acid derivatives against secretory phospholipase A2, a key enzyme involved in the inflammatory pathway.
Molecular bioSystems (2015-05-01)
K V Dileep, C Remya, J Cerezo, A Fassihi, H Pérez-Sánchez, C Sadasivan
ABSTRAKT
Inflammation is considered to be a key factor in major diseases like cancer, Alzheimer's disease, Parkinson's disease, etc. For the past few decades, pharmaceutical companies have explored new effective medications against inflammation. As a part of their detailed studies, many drug targets and drugs have been introduced against inflammation. In the present study, the inhibiting capacities of selected benzoic acid derivatives like gallic acid, vannilic acid, syringic acid and protocatechuic acid against secretory phospholipase A2 (sPLA2), a major enzyme involved in the inflammatory pathway, have been investigated. The detailed in vitro, biophysical and in silico studies carried out on these benzoic acid derivatives revealed that all the selected compounds have a uniform mode of binding in the active site of sPLA2 and are inhibitory in micromolar concentrations. The study also focuses on the non-selective inhibitory activity of an NSAID, aspirin, against sPLA2.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG