Merck
Agrimonia eupatoria L. and Cynara cardunculus L. Water Infusions: Phenolic Profile and Comparison of Antioxidant Activities.
Molecules (Basel, Switzerland) (2015-11-26)
Anika Kuczmannová, Peter Gál, Lenka Varinská, Jakub Treml, Ivan Kováč, Martin Novotný, Tomáš Vasilenko, Stefano Dall'Acqua, Milan Nagy, Pavel Mučaji
ABSTRAKT
Reactive oxygen species (ROS) are highly considered in the ethiopathogenesis of different pathological conditions because they may cause significant damage to cells and tissues. In this paper, we focused on potential antioxidant properties of two medical plants such as the Agrimonia eupatoria L. and Cynara cardunculus L. Both plants have previously been studied for their pharmacological activities, especially as hepatoprotective and hypoglycemic activities. It has been suggested, that their effects are related to the antioxidant properties of polyphenols, which are dominant compounds of the plants' extracts. In the present study HPLC-MS analysis of water infusion was performed allowing the identification of several phenolic constituents. Furthermore, antioxidant effects of the two extracts were compared showing higher effects for agrimony extract compared to artichoke. Thus, agrimony was selected for the in vivo study using the skin flap viability model. In conclusion, our results provide evidence that the A. eupatoria extract may be a valuable source of polyphenols to be studied for the future development of supplements useful in the prevention of diseases linked to oxidative stress.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)