Merck
RIP1-Dependent Programmed Necrosis is Negatively Regulated by Caspases During Hepatic Ischemia-Reperfusion.
Shock (Augusta, Ga.) (2015-05-27)
Dirk Rosentreter, Dominik Funken, Jörg Reifart, Konstantin Mende, Markus Rentsch, Andrej Khandoga
ABSTRAKT
Programmed necrosis (necroptosis), a newly discovered form of cell death, is mediated by receptor-interacting protein 1 (RIP1) and plays a pivotal role after myocardial, renal, and cerebral ischemia-reperfusion (I/R). The relevance of necroptosis in the postischemic liver remains, however, unclear. The aim of this study was to analyze the role of programed necrosis during hepatic I/R. C57BL6 mice were subjected to warm hepatic I/R (90 min/240 min). The animals were pretreated with either the RIP1 inhibitor necrostatin-1 (Nec-1, 3.5 μg kg) or vehicle (Nec-1inactive, 3.5 μg kg) administered systemically before ischemia. Sham-operated animals served as controls (n = 6 each group). The inflammatory response was evaluated by intravital microscopy. The hepatic transaminases alanine aminotransferase/aspartate aminotransferase in plasma as well as the activity of caspase-3 in tissue were determined as markers of hepatocellular injury. Leukocyte recruitment to the liver, sinusoidal perfusion failure, as well as the transaminase activities were strongly increased on I/R as compared with the sham-operated mice. Inhibition of the RIP1-dependent pathway with Nec-1, however, did not attenuate I/R-induced leukocyte migration, perfusion failure, and hepatocellular injury. Western blot analysis showed a baseline RIP1 expression in livers from sham-operated mice, whereas RIP1 expression was not detectable in both Nec-1-treated and vehicle-treated I/R group. Caspase-3 activity was significantly elevated after I/R in both postischemic groups. Our in vivo data show that RIP1-mediated necroptosis is not present in the postischemic liver and that I/R-induced caspase activation is associated with loss of RIP1 expression. Because caspases are able to cleave RIP1, we hypothesize that I/R-triggered caspase activation negatively regulates necroptosis and, thereby, determines apoptosis as a preferred route of cell death after hepatic I/R.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Fluorescein, free acid