Merck
Aging effect on the leaching behavior of heavy metals (Cu, Zn, and Cd) in red paddy soil.
Environmental science and pollution research international (2015-03-31)
Bin Huang, Zhongwu Li, Jinquan Huang, Guiqiu Chen, Xiaodong Nie, Wenming Ma, Hongbo Yao, Jiamei Zhen, Guangming Zeng
ABSTRAKT
Aging effect can influence the fractions distribution and mobility of metals after they are added into soil. In this study, incubation and soil column experiments under simulated acid rain condition were conducted to evaluate aging effect on the leaching characteristic of Cu, Zn, and Cd in artificial polluted red paddy soil. Our results showed that aging effect reduced metal contents in exchangeable and HoAc soluble fractions. Power function was the most excellent to describe the variation of exchangeable fraction, while pseudo first- and second-order functions were more successful to describe the leaching characteristic of metals from soil columns. The leaching amount of the metals from the polluted soil only accounted for a small part of their total content in soil, and the leachability of Cu was the weakest. Both the exchangeable and HoAc soluble fraction were available as indicators to evaluate the leachability of metals in red paddy soil. The shorter time the soil was contaminated, the more amounts of metals released from the soil. The reduction of exchangeable fraction caused by aging effect was the main reason for the decrease of metal mobility in soil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis