Merck
Genetic architecture of natural variation in Drosophila melanogaster aggressive behavior.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-24)
John Shorter, Charlene Couch, Wen Huang, Mary Anna Carbone, Jason Peiffer, Robert R H Anholt, Trudy F C Mackay
ABSTRAKT
Aggression is an evolutionarily conserved complex behavior essential for survival and the organization of social hierarchies. With the exception of genetic variants associated with bioamine signaling, which have been implicated in aggression in many species, the genetic basis of natural variation in aggression is largely unknown. Drosophila melanogaster is a favorable model system for exploring the genetic basis of natural variation in aggression. Here, we performed genome-wide association analyses using the inbred, sequenced lines of the Drosophila melanogaster Genetic Reference Panel (DGRP) and replicate advanced intercross populations derived from the most and least aggressive DGRP lines. We identified genes that have been previously implicated in aggressive behavior as well as many novel loci, including gustatory receptor 63a (Gr63a), which encodes a subunit of the receptor for CO2, and genes associated with development and function of the nervous system. Although genes from the two association analyses were largely nonoverlapping, they mapped onto a genetic interaction network inferred from an analysis of pairwise epistasis in the DGRP. We used mutations and RNAi knock-down alleles to functionally validate 79% of the candidate genes and 75% of the candidate epistatic interactions tested. Epistasis for aggressive behavior causes cryptic genetic variation in the DGRP that is revealed by changing allele frequencies in the outbred populations derived from extreme DGRP lines. This phenomenon may pertain to other fitness traits and species, with implications for evolution, applied breeding, and human genetics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Autocamtide 2-related inhibitory peptide, ≥97% (HPLC), lyophilized powder
Sigma-Aldrich
Aluminum isopropoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum isopropoxide, ≥98%