Merck
  • Development of a magnetic solid-phase extraction coupled with high-performance liquid chromatography method for the analysis of polyaromatic hydrocarbons.
Development of a magnetic solid-phase extraction coupled with high-performance liquid chromatography method for the analysis of polyaromatic hydrocarbons.
Journal of separation science (2015-05-02)
Yan Ma, Jiawen Xie, Jing Jin, Wei Wang, Zhijian Yao, Qing Zhou, Aimin Li, Ying Liang
ABSTRAKT
A novel magnetic solid phase extraction coupled with high-performance liquid chromatography method was established to analyze polyaromatic hydrocarbons in environmental water samples. The extraction conditions, including the amount of extraction agent, extraction time, pH and the surface structure of the magnetic extraction agent, were optimized. The results showed that the amount of extraction agent and extraction time significantly influenced the extraction performance. The increase in the specific surface area, the enlargement of pore size, and the reduction of particle size could enhance the extraction performance of the magnetic microsphere. The optimized magnetic extraction agent possessed a high surface area of 1311 m(2) /g, a large pore size of 6-9 nm, and a small particle size of 6-9 μm. The limit of detection for phenanthrene and benzo[g,h,i]perylene in the developed analysis method was 3.2 and 10.5 ng/L, respectively. When applied to river water samples, the spiked recovery of phenanthrene and benzo[g,h,i]perylene ranged from 89.5-98.6% and 82.9-89.1%, respectively. Phenanthrene was detected over a concentration range of 89-117 ng/L in three water samples withdrawn from the midstream of the Huai River, and benzo[g,h,i]perylene was below the detection limit.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG