Merck
  • Accuracy and feasibility of estimated tumour volumetry in primary gastric gastrointestinal stromal tumours: validation using semiautomated technique in 127 patients.
Accuracy and feasibility of estimated tumour volumetry in primary gastric gastrointestinal stromal tumours: validation using semiautomated technique in 127 patients.
European radiology (2015-05-21)
Sree Harsha Tirumani, Atul B Shinagare, Ailbhe C O'Neill, Mizuki Nishino, Michael H Rosenthal, Nikhil H Ramaiya
ABSTRAKT
To validate estimated tumour volumetry in primary gastric gastrointestinal stromal tumours (GISTs) using semiautomated volumetry. In this IRB-approved retrospective study, we measured the three longest diameters in x, y, z axes on CTs of primary gastric GISTs in 127 consecutive patients (52 women, 75 men, mean age 61 years) at our institute between 2000 and 2013. Segmented volumes (Vsegmented) were obtained using commercial software by two radiologists. Estimate volumes (V1-V6) were obtained using formulae for spheres and ellipsoids. Intra- and interobserver agreement of Vsegmented and agreement of V1-6 with Vsegmented were analysed with concordance correlation coefficients (CCC) and Bland-Altman plots. Median Vsegmented and V1-V6 were 75.9, 124.9, 111.6, 94.0, 94.4, 61.7 and 80.3 cm(3), respectively. There was strong intra- and interobserver agreement for Vsegmented. Agreement with Vsegmented was highest for V6 (scalene ellipsoid, x ≠ y ≠ z), with CCC of 0.96 [95 % CI 0.95-0.97]. Mean relative difference was smallest for V6 (0.6 %), while it was -19.1 % for V5, +14.5 % for V4, +17.9 % for V3, +32.6 % for V2 and +47 % for V1. Ellipsoidal approximations of volume using three measured axes may be used to closely estimate Vsegmented when semiautomated techniques are unavailable. Estimation of tumour volume in primary GIST using mathematical formulae is feasible. Gastric GISTs are rarely spherical. Segmented volumes are highly concordant with three axis-based scalene ellipsoid volumes. Ellipsoid volume can be used as an alternative for automated tumour volumetry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)