Merck
Bivalirudin vs Heparin in Patients Who Undergo Transcatheter Aortic Valve Implantation.
The Canadian journal of cardiology (2015-07-28)
Philipp Lange, Martin Greif, Dario Bongiovanni, Antonia Thaumann, Michael Näbauer, Bernhard Bischoff, Susanne Helbig, Christoph Becker, Christoph Schmitz, Melvin D'Anastasi, Julinda Mehilli, Peter Boekstegers, Steffen Massberg, Christian Kupatt
ABSTRAKT
We aimed to compare safety and efficacy of the direct thrombin inhibitor bivalirudin with unfractionated heparin (UFH) during transcatheter aortic valve implantation (TAVI). In this retrospective analysis, 461 patients underwent TAVI between 2007 and 2012; 339 patients received bivalirudin, and 122 patients received UFH. In the bivalirudin group, the Sapien XT valve was implanted in 159 (46.9%) patients, and 180 (53.1%) received a Medtronic CoreValve. In the UFH group, only the Medtronic CoreValve was implanted. The primary outcome of interest was the incidence of any bleeding. Secondary outcomes of interest were all-cause mortality and cardiovascular mortality at 72 hours after the procedure and at 30 days. No significant difference between the groups was observed for life-threatening bleeding (2.4% for bivalirudin vs 3.3% for UFH; P = 0.59), major bleeding (8.3% vs 8.2%, respectively; P = 0.98) and minor bleeding (8.3% vs 7.4%, respectively; P = 0.76). At 72 hours after the procedure, all-cause mortality was 3.0% in the bivalirudin group and 3.3% for the UFH group (P = 0.88), whereas cardiovascular mortality was 3.0% in the bivalirudin group and 2.5% in the heparin group (P = 0.77). At 30 days, all-cause mortality was 5.3% vs 4.1% in the bivalirudin and heparin groups (P = 0.57) and cardiovascular mortality was 4.4% vs 2.5% (P = 0.33). Device success (Valve Academic Research Consortium 2 composite end point) was 94.0% in the bivalirudin-treated and 92.6% in the UFH-treated patients (P = 0.60). The early safety at 30 days was 85.3% in the bivalirudin-treated group compared with 83.6% in the UFH-treated group (P = 0.65). Bivalirudin has a safety and efficacy profile similar to weight-adjusted UFH during the TAVI procedure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bivalirudin trifluoroacetate salt, ≥97% (HPLC)