Merck
Oral versus intravenous paracetamol: which is better in closure of patent ductus arteriosus in very low birth weight infants?
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians (2014-12-05)
Selim Sancak, Tulin Gokmen Yildirim, Sevilay Topcuoglu, Taner Yavuz, Guner Karatekin, Fahri Ovali
ABSTRAKT
To compare the efficacy of oral and intravenous paracetamol for closure of hemodynamically significant patent ductus arteriosus (HSPDA) in very low birth weight (VLBW) preterm infants. Eighteen VLBW infants with HSPDA treated with either intravenous (n = 10) or oral (n = 8) paracetamol at 60 mg/kg/d for three consecutive days were analysed retrospectively. Ductal closure rate and evaluation of liver function tests were the major outcomes. After two courses of treatment, HSPDA closure rate was higher in oral paracetamol group than that in the intravenous paracetamol group (88% versus 70%), but it was not statistically significant (p = 0.588). Liver function tests were normal after the treatment. Although it was not statistically significant, the cumulative closure rates were higher in oral paracetamol group than those in the intravenous group. Larger trials are needed to confirm these data.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®