Merck
Exciton Fine Structure of CdSe/CdS Nanocrystals Determined by Polarization Microscopy at Room Temperature.
ACS nano (2015-07-28)
Stefano Vezzoli, Mathieu Manceau, Godefroy Leménager, Quentin Glorieux, Elisabeth Giacobino, Luigi Carbone, Massimo De Vittorio, Alberto Bramati
ABSTRAKT
We present a method that allows determining the band-edge exciton fine structure of CdSe/CdS dot-in-rods samples based on single particle polarization measurements at room temperature. We model the measured emission polarization of such single particles considering the fine structure properties, the dielectric effect induced by the anisotropic shell, and the measurement configuration. We use this method to characterize the band-edge exciton fine structure splitting of various samples of dot-in-rods. We show that, when the diameter of the CdSe core increases, a transition from a spherical like band-edge exciton symmetry to a rod-like band edge exciton symmetry occurs. This explains the often reported large emission polarization of such particles compared to spherical CdSe/CdS emitters.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Octadecylphosphonic acid, 97%
Sigma-Aldrich
Cadmium oxide, powder, 99.5% trace metals basis
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis