Merck
Species identification within Acinetobacter calcoaceticus-baumannii complex using MALDI-TOF MS.
Journal of microbiological methods (2015-09-19)
Benjamin E W Toh, David L Paterson, Witchuda Kamolvit, Hosam Zowawi, David Kvaskoff, Hanna Sidjabat, Alexander Wailan, Anton Y Peleg, Charlotte A Huber
ABSTRAKT
Acinetobacter baumannii, one of the more clinically relevant species in the Acinetobacter genus is well known to be multi-drug resistant and associated with bacteremia, urinary tract infection, pneumonia, wound infection and meningitis. However, it cannot be differentiated from closely related species such as Acinetobacter calcoaceticus, Acinetobacter pittii and Acinetobacter nosocomialis by most phenotypic tests and can only be differentiated by specific, time consuming genotypic tests with very limited use in clinical microbiological laboratories. As a result, these species are grouped into the A. calcoaceticus-A. baumannii (Acb) complex. Herein we investigated the mass spectra of 73 Acinetobacter spp., representing ten different species, using an AB SCIEX 5800 MALDI-TOF MS to differentiate members of the Acinetobacter genus, including the species of the Acb complex. RpoB gene sequencing, 16S rRNA sequencing, and gyrB multiplex PCR were also evaluated as orthogonal methods to identify the organisms used in this study. We found that whilst 16S rRNA and rpoB gene sequencing could not differentiate A. pittii or A. calcoaceticus, they can be differentiated using gyrB multiplex PCR and MALDI-TOF MS. All ten Acinetobacter species investigated could be differentiated by their MALDI-TOF mass spectra.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)